Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcm3ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019434.2 | NP_062307.2 | 28 | 6017 | ||
GenBank mRNA | AJ006590.1 | - | 28 | 6050 | ||
AK122313.1 | - | 28 | 6061 | |||
AK165254.1 | - | 17 | 2722 | |||
BC052452.1 | - | 28 | 6017 | 6409827 | ||
BC067414.1 | - | 27 | 4952 |