Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncapg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019438.1 | NP_062311.1 | 3, 4 | 638 | ||
GenBank mRNA | AK017756.1 | - | 3, 4 | 533 | ||
CJ068152.1 | - | 3, 4 | 639 | |||
CX564513.1 | - | 2, 3 | 283 |