Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncapg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019438.1 | NP_062311.1 | 5 | 849 | ||
GenBank mRNA | CX564513.1 | - | 4 | 494 |