Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019442.3 | NP_062315.1 | 2, 3 | 262 | ||
GenBank mRNA | AF099934.1 | - | 2, 3 | 196 | ||
AK146739.1 | - | 2, 3 | 264 | |||
AK168865.1 | - | 2, 3 | 264 | |||
AK215314.1 | - | 1, 2 | 22 | |||
AK218509.1 | - | 1, 2 | 91 | |||
BC022681.1 | - | 2, 3 | 262 | 4240347 |