Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc4a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136260.1 | NP_001129732.1 | 14, 15 | 1970 | ||
NM_001197147.1 | NP_001184076.1 | 14, 15 | 1943 | |||
NM_018760.2 | NP_061230.2 | 14, 15 | 1970 | |||
GenBank mRNA | AF020195.1 | - | 14, 15 | 1933 | ||
AF141934.3 | - | 11, 12 | 1772 | |||
BC026592.1 | - | 5, 6 | 763 | |||
BC129863.1 | - | 14, 15 | 1932 | |||
BC129864.1 | - | 14, 15 | 1931 | |||
BC148292.2 | - | 14, 15 | 1906 | 40110074 | ||
HQ018820.1 | - | 11, 12 | 1807 | |||
HQ204220.1 | - | 14, 15 | 1914 | |||
HQ285250.1 | - | 11, 12 | 1783 |