Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019445.2 | NP_062318.2 | 2 | 1859 | ||
GenBank mRNA | AF218940.1 | - | 2 | 1854 | ||
BC094606.1 | - | 2 | 1859 | 6840339 |