Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apbb1ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019456.2 | NP_062329.2 | 13 | 1670 | ||
GenBank mRNA | AF020313.1 | - | 10 | 1274 | ||
AK089987.1 | - | 5 | 544 | |||
BC003991.1 | - | 10 | 1321 | |||
BC023110.1 | - | 13 | 1559 | 3707478 |