Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apbb1ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019456.2 | NP_062329.2 | 4 | 545 | ||
GenBank mRNA | AF020313.1 | - | 1 | 157 | ||
AK021331.1 | - | 5 | 424 | |||
AK040084.1 | - | 4 | 480 | |||
AK041552.1 | - | 4 | 548 | |||
AK042467.1 | - | 4 | 483 | |||
AK079736.1 | - | 4 | 779 | |||
AK162323.1 | - | 4 | 429 | |||
BB638237.1 | - | 4 | 547 | |||
BC023110.1 | - | 4 | 440 | 3707478 |