Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nap1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138742.1 | NP_620081.1 | 1 | 1199 | ||
GenBank mRNA | AB010711.1 | - | 1 | 1215 | ||
AK078521.1 | - | 1 | 1191 | |||
BC020176.1 | - | 1 | 1199 | 4505496 | ||
BC027045.1 | - | 1 | 1096 | 5363560 |