Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019457.2 | NP_062330.1 | 5 | 747 | ||
GenBank mRNA | AF092208.1 | - | 5 | 692 | ||
AK016236.1 | - | 5 | 736 | |||
AK077124.1 | - | 5 | 731 | |||
BC046277.1 | - | 5 | 698 | 6310345 |