Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Socs6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018821.4 | NP_061291.2 | 3 | 1230 | ||
GenBank mRNA | AF121907.1 | - | 2 | 1137 | ||
AK028665.1 | - | 3 | 1224 | |||
AK028701.1 | - | 3 | 1216 | |||
AK031164.1 | - | 3 | 1231 | |||
AK044946.1 | - | 3 | 1226 | |||
AK083341.1 | - | 3 | 1215 | |||
AK142069.1 | - | 3 | 1346 | |||
AK156591.1 | - | 4 | 1437 | |||
AK217338.1 | - | 1 | 170 | |||
BC017597.1 | - | 2 | 1132 | 4482828 | ||
BC085245.1 | - | 4 | 1058 | 30444479 | ||
BC094443.1 | - | 4 | 1285 | 4017254 |