Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubqln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018798.2 | NP_061268.2 | 1 | 830 | ||
GenBank mRNA | AF177346.1 | - | 1 | 831 | ||
AK076159.1 | - | 1 | 834 | |||
BC021824.1 | - | 1 | 215 | |||
BC053022.1 | - | 1 | 830 | 5708544 |