Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpf40b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018786.2 | NP_061256.1 | 10 | 639 | ||
GenBank mRNA | AF135440.1 | - | 10 | 639 | ||
AK140075.1 | - | 10 | 808 | |||
AK164947.1 | - | 11 | 852 | |||
BC051961.1 | - | 9 | 618 | |||
BC058389.1 | - | 8 | 1543 | |||
BC082994.1 | - | 10 | 653 | 6333535 |