Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Extl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018788.3 | NP_061258.2 | 3 | 2428 | ||
GenBank mRNA | AF083550.1 | - | 1 | 2218 | ||
AK137842.1 | - | 1 | 1505 | |||
AK220344.1 | - | 1 | 1771 | |||
BC058526.1 | - | 1 | 1819 | |||
BC065073.1 | - | 1 | 1893 | 6834580 |