Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019458.3 | NP_062331.2 | 6 | 659 | ||
GenBank mRNA | AB041615.1 | - | 6 | 677 | ||
AK017762.1 | - | 6 | 729 | |||
AK134857.1 | - | 6 | 672 | |||
AK148538.1 | - | 6 | 694 | |||
AK151665.1 | - | 6 | 705 | |||
AK152778.1 | - | 6 | 675 | |||
AK152836.1 | - | 6 | 705 | |||
BC010317.1 | - | 5 | 703 | |||
BC029843.1 | - | 6 | 659 | 2651081 | ||
BC083337.1 | - | 6 | 640 | 5701517 |