Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gripap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290455.1 | NP_001277384.1 | 22 | 1966 | ||
NM_207670.2 | NP_997553.1 | 21 | 1873 | |||
GenBank mRNA | AK156165.1 | - | 21 | 1874 | ||
AK172623.1 | - | 21 | 1874 | |||
AK172699.1 | - | 21 | 1874 | |||
AK173119.1 | - | 9 | 889 | |||
AW342118.1 | - | 21 | 1848 | |||
BC138523.1 | - | 22 | 1962 | 8861543 | ||
BC145310.1 | - | 22 | 1962 | 9053864 |