Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp8b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081182.2 | NP_001074651.2 | 21 | 2425 | ||
GenBank mRNA | AK155736.1 | - | 20 | 2459 | ||
AK173107.1 | - | 10 | 1870 |