Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp8b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001488.3 | NP_001001488.2 | 15 | 1935 | ||
GenBank mRNA | AK036959.1 | - | 15 | 1857 | ||
AK157316.1 | - | 15 | 2203 | |||
AY506548.1 | - | 15 | 1664 | |||
BC117946.2 | - | 15 | 1662 | 40098035 |