Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3glb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282037.1 | NP_001268966.1 | 10 | 1162 | ||
NM_001282042.1 | NP_001268971.1 | 8 | 1099 | |||
NM_019464.3 | NP_062337.1 | 8 | 1099 | |||
GenBank mRNA | AF263364.1 | - | 8 | 1087 | ||
AF272946.1 | - | 10 | 1150 | |||
AK004590.1 | - | 8 | 910 | |||
AK029647.1 | - | 7 | 760 | |||
AK146246.1 | - | 8 | 928 | |||
AK158415.1 | - | 8 | 875 | |||
AK172959.1 | - | 8 | 910 | |||
AK184553.1 | - | 2 | 146 | |||
BC024362.1 | - | 8 | 909 | 5031296 | ||
DQ205686.1 | - | 8 | 1087 |