Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fezf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080433.3 | NP_536681.2 | 4, 5 | 1447 | ||
GenBank mRNA | AB042399.1 | - | 3, 4 | 1164 | ||
AI325906.1 | - | 1, 2 | 97 | |||
AK019996.1 | - | 4, 5 | 1411 | |||
BC055718.1 | - | 3, 4 | 1774 | 6415359 |