Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tyk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205312.1 | NP_001192241.1 | 17 | 2734 | ||
NM_018793.2 | NP_061263.2 | 17 | 2665 | |||
GenBank mRNA | AF173032.1 | - | 17 | 2600 | ||
AK154438.1 | - | 17 | 2692 | |||
AK155923.1 | - | 17 | 2577 | |||
AK159303.1 | - | 17 | 2634 | |||
AK167726.1 | - | 17 | 2644 | |||
AK170107.1 | - | 17 | 2604 | |||
AK170940.1 | - | 17 | 2603 | |||
AK171609.1 | - | 17 | 2577 | |||
AK206664.1 | - | 2 | 195 | |||
BC019789.1 | - | 1 | 81 | |||
BC053021.1 | - | 11 | 2305 | |||
BC094240.1 | - | 16 | 2561 | 30610201 |