Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cadm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025600.1 | NP_001020771.1 | 3, 4 | 453 | ||
NM_018770.3 | NP_061240.3 | 3, 4 | 453 | |||
NM_207675.2 | NP_997558.2 | 3, 4 | 453 | |||
NM_207676.2 | NP_997559.1 | 3, 4 | 453 | |||
GenBank mRNA | AB052293.1 | - | 3, 4 | 453 | ||
AB064265.2 | - | 3, 4 | 432 | |||
AB092414.1 | - | 3, 4 | 432 | |||
AB183399.1 | - | 3, 4 | 432 | |||
AB183400.1 | - | 3, 4 | 432 | |||
AB183401.1 | - | 3, 4 | 432 | |||
AB183402.1 | - | 3, 4 | 432 | |||
AB377192.1 | - | 3, 4 | 453 | |||
AF061260.1 | - | 2, 3 | 290 | |||
AF434663.1 | - | 3, 4 | 548 | |||
AF539424.1 | - | 3, 4 | 445 | |||
AK013775.1 | - | 3, 4 | 454 | |||
AK013911.1 | - | 3, 4 | 547 | |||
AK053839.1 | - | 3, 4 | 454 | |||
AY351388.1 | - | 3, 4 | 432 | |||
BC057125.1 | - | 3, 4 | 441 | 6414884 | ||
BC090970.1 | - | 3, 4 | 632 | 6703838 | ||
BC095986.1 | - | 3, 4 | 445 | 5372277 | ||
DQ279856.1 | - | 3, 4 | 432 | |||
JQ782219.1 | - | 3, 4 | 432 |