Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289492.1 | NP_001276421.1 | 7 | 1108 | ||
NM_001289493.1 | NP_001276422.1 | 6 | 933 | |||
NM_018734.3 | NP_061204.3 | 6 | 970 | |||
GenBank mRNA | AK132266.1 | - | 6 | 969 | ||
BC019195.1 | - | 7 | 1019 | 5036920 | ||
U44731.1 | - | 6 | 822 |