Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc13a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019481.2 | NP_062354.2 | 8, 9 | 952 | ||
GenBank mRNA | AF199365.1 | - | 8, 9 | 952 | ||
AF199366.1 | - | 8, 9 | 952 | |||
AK005153.1 | - | 2, 3 | 287 | |||
AK128899.1 | - | 8, 9 | 952 | |||
BC022672.1 | - | 4, 5 | 331 | |||
BC040789.1 | - | 8, 9 | 926 | |||
BC049981.1 | - | 8, 9 | 934 | 5065026 |