Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Impa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018864.5 | NP_061352.2 | 7, 8 | 692 | ||
GenBank mRNA | AF042730.1 | - | 7, 8 | 637 | ||
AK011770.1 | - | 7, 8 | 630 | |||
AK078406.1 | - | 7, 8 | 641 | |||
AK083238.1 | - | 7, 8 | 626 | |||
AK145472.1 | - | 629 | ||||
AK147778.1 | - | 7, 8 | 652 | |||
AK152774.1 | - | 7, 8 | 643 | |||
AK153457.1 | - | 7, 8 | 659 | |||
AK160203.1 | - | 7, 8 | 630 | |||
AK160467.1 | - | 7, 8 | 624 | |||
AK165977.1 | - | 7, 8 | 634 | |||
AK168457.1 | - | 7, 8 | 667 | |||
AK210121.1 | - | 2, 3 | 177 | |||
BC049080.1 | - | 7, 8 | 633 | 6419835 | ||
BC055722.1 | - | 6, 7 | 587 | 6413389 |