Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018881.3 | NP_061369.2 | 6 | 710 | ||
GenBank mRNA | AF184981.1 | - | 5 | 775 | ||
AK129026.1 | - | 2 | 243 | |||
BC031415.1 | - | 6 | 710 | 3990305 |