Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285870.1 | NP_001272799.1 | 7 | 1682 | ||
NM_001285871.1 | NP_001272800.1 | 7 | 1867 | |||
NM_001285873.1 | NP_001272802.1 | 7 | 1682 | |||
NM_018880.3 | NP_061368.1 | 7 | 1656 | |||
GenBank mRNA | AB030912.1 | - | 7 | 1683 | ||
AF220019.1 | - | 7 | 1575 | |||
AK019165.1 | - | 7 | 1617 | |||
AK144674.1 | - | 7 | 1585 | |||
AK170029.1 | - | 7 | 1620 | |||
AK202039.1 | - | 2 | 139 | |||
AK203347.1 | - | 2 | 138 | |||
AK204820.1 | - | 2 | 139 | |||
BC003347.1 | - | 7 | 1696 | |||
BC034263.1 | - | 7 | 1621 | 4503884 |