Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019488.4 | NP_062361.1 | 8 | 1039 | ||
GenBank mRNA | AF232061.2 | - | 8 | 996 | ||
AJ245936.1 | - | 8 | 1016 | |||
AK081806.1 | - | 8 | 1056 | |||
AK141193.1 | - | 5 | 1749 | |||
AK155792.1 | - | 8 | 1068 | |||
AK170319.1 | - | 8 | 1052 | |||
AY856043.1 | - | 8 | 1000 | |||
AY856044.1 | - | 7 | 891 | |||
AY856046.1 | - | 4 | 421 | |||
BC021758.1 | - | 4 | 365 | |||
BC090993.1 | - | 8 | 1039 | 6751763 | ||
Y17802.1 | - | 8 | 1021 |