Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207678.1 | NP_997561.1 | 9, 10 | 1143 | ||
GenBank mRNA | AB041605.1 | - | 9, 10 | 1120 | ||
AK048244.1 | - | 9, 10 | 2923 | |||
AY337018.1 | - | 9, 10 | 1126 | |||
BC003773.1 | - | 2, 3 | 266 | |||
BC023747.1 | - | 9, 10 | 2911 | |||
BC083055.1 | - | 2, 3 | 130 | |||
BC132295.1 | - | 9, 10 | 1135 | 40130572 | ||
U37351.1 | - | 2, 3 | 149 |