Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uqcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018888.4 | NP_061376.2 | 6 | 689 | ||
GenBank mRNA | AF253516.1 | - | 6 | 436 | ||
AK010376.1 | - | 6 | 430 | |||
AK011152.1 | - | 6 | 443 | |||
AK011953.1 | - | 6 | 411 | |||
AK077323.1 | - | 6 | 427 | |||
AK139488.1 | - | 6 | 689 | |||
AK169413.1 | - | 6 | 426 | |||
BC050884.1 | - | 6 | 642 | 6808756 | ||
BC057570.1 | - | 6 | 642 | 6808756 |