Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtpbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145979.1 | NP_001139451.1 | 3 | 326 | ||
NM_019581.3 | NP_062527.2 | 3 | 361 | |||
GenBank mRNA | AF168991.1 | - | 3 | 266 | ||
AK087511.1 | - | 3 | 232 | |||
AK146415.1 | - | 3 | 311 | |||
AK148020.1 | - | 3 | 282 | |||
BC049089.1 | - | 3 | 322 | 5720710 | ||
BC051958.1 | - | 3 | 322 | 5720710 | ||
BY240789.1 | - | 3 | 364 | |||
BY266706.1 | - | 3 | 327 |