Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019501.3 | NP_062374.2 | 10 | 1138 | ||
GenBank mRNA | AB210841.1 | - | 10 | 1058 | ||
AF118855.1 | - | 8 | 833 | |||
AK011869.1 | - | 1 | 1456 | |||
BC026820.1 | - | 10 | 1040 | 4039464 | ||
BC107273.1 | - | 10 | 953 | |||
BC107274.1 | - | 10 | 957 |