Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019537.2 | NP_062410.1 | 4, 5 | 461 | ||
GenBank mRNA | AJ238270.1 | - | 4, 5 | 463 | ||
AK042203.1 | - | 4, 5 | 472 | |||
AK088118.1 | - | 4, 5 | 474 | |||
BC032965.1 | - | 4, 5 | 454 | 1196077 | ||
BC060285.1 | - | 4, 5 | 475 | 6508084 |