Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tro | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002272.3 | NP_001002272.1 | 3, 4 | 1788 | ||
NM_001290770.1 | NP_001277699.1 | 3, 4 | 1737 | |||
NM_001290771.1 | NP_001277700.1 | 3, 4 | 1788 | |||
NM_001290772.1 | NP_001277701.1 | 3, 4 | 1788 | |||
NM_019548.4 | NP_062421.2 | 3, 4 | 1788 | |||
GenBank mRNA | AB017108.1 | - | 1, 2 | 129 | ||
AB032477.1 | - | 3, 4 | 1656 | |||
AB036783.1 | - | 4, 5 | 1730 | |||
AF241244.1 | - | 3, 4 | 1652 | |||
AF241245.1 | - | 3, 4 | 1652 | |||
AF288605.1 | - | 4, 5 | 1730 | |||
AF288606.1 | - | 4, 5 | 1730 | |||
AF319977.1 | - | 2, 3 | 1671 | |||
AK122449.1 | - | 2, 3 | 678 | |||
BC075630.1 | - | 3, 4 | 1737 | 30536880 |