Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpf40a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018785.2 | NP_061255.1 | 7, 8 | 664 | ||
GenBank mRNA | AF135439.1 | - | 7, 8 | 664 | ||
AK087963.1 | - | 7, 8 | 581 | |||
BC003318.1 | - | 7, 8 | 691 |