Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019550.2 | NP_062423.1 | 4 | 387 | ||
GenBank mRNA | AF095718.2 | - | 4 | 257 | ||
AK041655.1 | - | 4 | 286 | |||
AK050624.1 | - | 4 | 268 | |||
AK050650.1 | - | 4 | 268 | |||
AK135738.1 | - | 4 | 388 | |||
AK137348.1 | - | 4 | 266 | |||
AK207246.1 | - | 4 | 218 | |||
BC010255.1 | - | 4 | 273 | 3709255 |