Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019550.2 | NP_062423.1 | 13 | 1626 | ||
GenBank mRNA | AF095718.2 | - | 13 | 1496 | ||
AK013734.1 | - | 2 | 189 | |||
AK033964.1 | - | 6 | 2033 | |||
AK135738.1 | - | 13 | 1629 | |||
AK137348.1 | - | 13 | 1505 | |||
BC010255.1 | - | 13 | 1512 | 3709255 |