Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019551.2 | NP_062424.1 | 5, 6 | 742 | ||
GenBank mRNA | AJ251328.1 | - | 5, 6 | 716 | ||
AK009089.1 | - | 5, 6 | 716 | |||
AK135925.1 | - | 5, 6 | 754 | |||
AK167707.1 | - | 4, 5 | 538 | |||
BC132511.1 | - | 5, 6 | 705 | 40130788 | ||
BC132513.1 | - | 5, 6 | 705 | 40130790 | ||
BC145334.1 | - | 4, 5 | 534 |