Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019552.2 | NP_062425.1 | 13 | 2406 | ||
GenBank mRNA | AF266284.1 | - | 13 | 2136 | ||
AK081782.1 | - | 13 | 2118 | |||
BC046818.1 | - | 13 | 2362 | 5702696 | ||
BC053020.1 | - | 13 | 2362 | 5702696 | ||
BC054793.1 | - | 13 | 2406 | 6833785 |