Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019553.2 | NP_062426.2 | 5 | 1111 | ||
GenBank mRNA | AF159131.3 | - | 5 | 1072 | ||
AK010281.1 | - | 5 | 991 | |||
AK145578.1 | - | 5 | 1095 | |||
AK160095.1 | - | 5 | 1085 | |||
BC030895.1 | - | 4 | 543 | |||
BC043655.1 | - | 5 | 1083 | 5363948 | ||
BC059237.1 | - | 5 | 1071 | 6412782 | ||
BC060220.1 | - | 5 | 1063 |