Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Becn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019584.3 | NP_062530.2 | 10, 11 | 1172 | ||
GenBank mRNA | AF077302.2 | - | 9, 10 | 1034 | ||
AK028851.1 | - | 8, 9 | 2614 | |||
AK031501.1 | - | 9, 10 | 1799 | |||
AK032176.1 | - | 1173 | ||||
AK050329.1 | - | 10, 11 | 1173 | |||
AK077392.1 | - | 10, 11 | 1259 | |||
AK083800.1 | - | 10, 11 | 1162 | |||
AK151531.1 | - | 10, 11 | 1139 | |||
AK215649.1 | - | 2, 3 | 204 | |||
BC005770.1 | - | 10, 11 | 1153 | 2650059 |