Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Extl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019578.2 | NP_062524.2 | 1 | 1108 | ||
GenBank mRNA | AF224461.1 | - | 1 | 340 | ||
AK085075.1 | - | 1 | 1110 | |||
AK161915.1 | - | 1 | 181 | |||
AK164184.1 | - | 1 | 1106 | |||
AK172619.1 | - | 1 | 1110 | |||
BC120890.1 | - | 1 | 340 | 40061413 | ||
BC120891.1 | - | 1 | 340 | 40061414 |