Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019873.2 | NP_063926.1 | 2 | 831 | ||
GenBank mRNA | AK139541.1 | - | 1 | 412 | ||
BC046338.1 | - | 2 | 831 | 6395929 |