Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acpp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019807.2 | NP_062781.2 | 8 | 886 | ||
NM_207668.2 | NP_997551.1 | 8 | 886 | |||
GenBank mRNA | AF210243.1 | - | 8 | 839 | ||
AK020693.1 | - | 8 | 848 | |||
AK029273.1 | - | 8 | 872 | |||
AK076383.1 | - | 8 | 893 | |||
BC139826.1 | - | 8 | 877 | 40142642 |