Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dbn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177371.1 | NP_001170842.1 | 13 | 1678 | ||
NM_001177372.1 | NP_001170843.1 | 12 | 1540 | |||
NM_019813.4 | NP_062787.2 | 12 | 1540 | |||
GenBank mRNA | AB064321.1 | - | 12 | 1987 | ||
AF187147.1 | - | 13 | 1668 | |||
AF187148.1 | - | 12 | 1530 | |||
AK132060.1 | - | 12 | 1526 | |||
AK162697.1 | - | 12 | 1527 | |||
BC006714.1 | - | 13 | 1488 | 3710497 |