Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019721.2 | NP_062695.2 | 10 | 1723 | ||
GenBank mRNA | AF135789.1 | - | 8 | 1286 | ||
AK009492.1 | - | 7 | 784 | |||
AK035061.1 | - | 9 | 1541 | |||
AK085189.1 | - | 10 | 1710 | |||
BC012526.1 | - | 10 | 1707 | 3981018 | ||
BE979768.1 | - | 2 | 309 |
Gene Symbol: Tox4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK137050.1 | - | 3289 |