Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Net1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047159.2 | NP_001040624.1 | 2, 3 | 395 | ||
NM_019671.3 | NP_062645.2 | 4, 5 | 516 | |||
GenBank mRNA | AF094520.1 | - | 4, 5 | 412 | ||
AJ010045.1 | - | 2, 3 | 358 | |||
AK082128.1 | - | 2, 3 | 392 | |||
AK088632.1 | - | 2, 3 | 359 | |||
AK139921.1 | - | 2, 3 | 359 | |||
BC004699.1 | - | 2, 3 | 369 | 3499258 | ||
CN702942.1 | - | 2, 3 | 256 |