Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019718.2 | NP_062692.1 | 5 | 472 | ||
GenBank mRNA | AF143241.1 | - | 4 | 429 | ||
AK048644.1 | - | 5 | 480 | |||
AK185320.1 | - | 3 | 225 | |||
AK192226.1 | - | 3 | 224 | |||
BC042941.1 | - | 5 | 472 | 6493571 |