Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ivd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019826.3 | NP_062800.1 | 1, 2 | 169 | ||
GenBank mRNA | AF225989.1 | - | 1, 2 | 148 | ||
AK005024.1 | - | 1, 2 | 188 | |||
AK017660.1 | - | 1, 2 | 155 | |||
AK077416.1 | - | 1, 2 | 175 | |||
BC018325.1 | - | 1, 2 | 159 | 4192286 | ||
BC027198.1 | - | 1, 2 | 169 | 4237982 |