Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285793.1 | NP_001272722.1 | 4, 5 | 998 | ||
NM_019804.4 | NP_062778.2 | 4, 5 | 931 | |||
GenBank mRNA | AF142672.1 | - | 4, 5 | 908 | ||
AF158746.1 | - | 4, 5 | 910 | |||
AK018658.1 | - | 4, 5 | 895 | |||
AK041692.1 | - | 4, 5 | 923 | |||
AK045602.1 | - | 4, 5 | 903 | |||
AK050005.1 | - | 4, 5 | 927 | |||
AK052013.1 | - | 4, 5 | 925 | |||
AK053603.1 | - | 4, 5 | 992 | |||
AK085368.1 | - | 4, 5 | 917 | |||
AK136637.1 | - | 4, 5 | 843 | |||
AK214305.1 | - | 1, 2 | 22 | |||
BC013492.1 | - | 4, 5 | 894 | 4218187 | ||
BC031115.1 | - | 4, 5 | 983 | 5026168 |